Learn More
Tocris Bioscience™ APETx2
GREENER_CHOICE
Catalog No. 4804100U
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
48-041-00U 100 μg
1 options
This item is not returnable. View return policy
Catalog No. 48-041-00U Supplier Tocris Bioscience™ Supplier No. 4804/100U
Add to Cart
Add to Cart
This item is not returnable. View return policy
ASIC3 channel blocker
APETx2 is an acid-sensing ion channel 3 (ASIC3) channel blocker (IC50 values are 63 and 175 nM for homomeric rat and human ASIC3 channels). Also inhibits NaV1.8 and NaV1.2 channels (IC50 values are 55 and 114 nM respectively). Demonstrates analgesic properties against acid-induced and inflammatory pain.
Specifications
Purity 0.95