Learn More
Tocris Bioscience™ [Arg8]-Vasopressin
GREENER_CHOICE
Catalog No. 29351
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
29-351 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 29-351 Supplier Tocris Bioscience™ Supplier No. 2935/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Antidiuretic hormone
[Arg8]-Vasopressin is an antidiuretic hormone found in most mammalian species. Also acts as a neurotransmitter at synapses in the brain; increases [Ca2+]i in cultured rat hippocampal neurons.
Specifications
Quantity 1 mg