Learn More
Tocris Bioscience™ Bax inhibitor peptide, negative control
Catalog No. 17871
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
17-871 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 17-871 Supplier Tocris Bioscience™ Supplier No. 1787/1
This item is not returnable. View return policy
Negative control peptide for Bax inhibitor peptide V5 (Cat. No. 1785)
Negative control peptide for the Bax inhibitor peptide V5, which inhibits Bax translocation to mitochondria and Bax-mediated apoptosis in vitro.
Active Analog V5 also available.
Specifications
Quantity 1 mg