Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ BAY 55-9837
GREENER_CHOICE

Catalog No. 27111
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-111 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 27-111 Supplier Tocris Bioscience™ Supplier No. 2711/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective VPAC2 agonist

Bay 55-9837 is a selective VPAC2 receptor agonist (EC50 values are 0.4, 100 and >1000 nM for VPAC2, VPAC1 and PAC1, respectively in a cAMP accumulation assay; IC50 values are 60, 8700 and >10000 nM for VPAC2, VPAC1 and PAC1, respectively in a competition binding assay). Stimulates glucose-dependent insulin secretion in isolated human pancreatic islets. Reduces HIV-1 viral replication and shows cooperative effects when given in conjunction with VPAC1 agonists.

Specifications

Purity 95%
Content And Storage Store at -20°C
Molecular Weight (g/mol) M.W. (Observed): 3742.29
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.