Learn More
Tocris Bioscience™ BIO 1211
GREENER_CHOICE
Catalog No. 39101
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
39-101 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 39-101 Supplier Tocris Bioscience™ Supplier No. 3910/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selective α4β1 (VLA-4) inhibitor
BIO 1211 is a selective, high affinity α4β1 (Very Late Antigen 4; VLA-4) inhibitor; displays 200-fold selectivity for the activated form of α4β1 (KD = 70 pM; IC50 = 0.004 μM). Selective for α4β1 over a range of other integrins (IC50 >100 μM for α1β1, α5β1 and α6β1).
Specifications
Purity 95%
Quantity 1 mg