Learn More
Tocris Bioscience™ Bradykinin
GREENER_CHOICE
Catalog No. 30045
Change view
Click to view available options
Quantity:
5 mg
Catalog No. Quantity
30-045 5 mg
1 options
This item is not returnable. View return policy
Catalog No. 30-045 Supplier Tocris Bioscience™ Supplier No. 3004/5
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Endogenous agonist at bradykinin receptors (B2 > B1)
Bradykinin is an endogenous bradykinin receptor agonist that displays selectivity for B2 over B1 receptors. Proinflammatory peptide.
Specifications
Quantity 5 mg