Learn More
Tocris Bioscience™ Brain natriuretic peptide (1-32) (human)
GREENER_CHOICE
Catalog No. 35221
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
35-221 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 35-221 Supplier Tocris Bioscience™ Supplier No. 3522/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Endogenous peptide agonist at ANP receptor A (NPR1)
Brain natriuretic peptide (1-32) (human) is an endogenous peptide secreted from cardiac ventricles in response to volume increase and pressure overload that acts as an agonist at atrial natriuretic peptide (ANP) receptor A (NRP1). Decreases de novo collagen synthesis and increases MMP gene expression in vitro. Exhibits natriuretic, vasodilatory and lusitropic activity and inhibits the sympathetic and renin-angiotensin-aldosterone systems in vivo.
Specifications
Purity 0.95
Quantity 1 mg