Learn More
Tocris Bioscience™ C-type natriuretic peptide (1-22) (human, rat, swine)
GREENER_CHOICE
Catalog No. 3520500U
Change view
Click to view available options
Quantity:
500 μg
Catalog No. Quantity
35-205-00U 500 μg
1 options
This item is not returnable. View return policy
Catalog No. 35-205-00U Supplier Tocris Bioscience™ Supplier No. 3520/500U
Add to Cart
Add to Cart
This item is not returnable. View return policy
Endogenous peptide agonist at NPR2
C-type natriuretic peptide (1-22) (human, rat, swine) is an endogenous peptide found in plasma and cerebrospinal fluid. Behaves as an agonist at natriuretic peptide receptor NPR2 (NPRB) and exhibits affinity for NPR3 (NPRC). Inhibits L-type calcium currents in myocytes and exhibits antiproliferative effects in cardiac fibroblasts in vitro. Regulates cartilage homeostasis, body fluid volume and exhibits vasodilatory activity in vivo.
Specifications
Quantity 500 μg