Learn More
Tocris Bioscience™ Caffeic acid-pYEEIE
Catalog No. 19351
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
19-351 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 19-351 Supplier Tocris Bioscience™ Supplier No. 1935/1
This item is not returnable. View return policy
Phosphopeptide ligand for src SH2 domain
Caffeic acid-pYEEIE is a phosphopeptide ligand for the src SH2 domain (IC50 = 42 nM); displays 30-fold higher affinity than N-acetyl-O-phosphono-Tyr-Glu-Glu-Ile-Glu ( Ac-pYEEIE, Cat. No. 1927).
Specifications
Quantity 1 mg