Learn More
Tocris Bioscience™ CART (55-102) (rat)
GREENER_CHOICE
Catalog No. 3337100U
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
33-371-00U 100 μg
1 options
This item is not returnable. View return policy
Catalog No. 33-371-00U Supplier Tocris Bioscience™ Supplier No. 3337/100U
Add to Cart
Add to Cart
This item is not returnable. View return policy
Neuromodulatory neuropeptide fragment; satiety factor
CART (55-102) (rat) is a cocaine- and amphetamine-regulated transcript (CART) with potent appetite-suppressing activity. Satiety factor; inhibits normal and starvation-induced feeding. Closely related to the actions of leptin and neuropeptide Y; blocks the neuropeptide Y-induced feeding response. Induces anxiety and stress behavior in rodents.
Specifications
Purity 95%