Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CCK Octapeptide, sulfated
GREENER_CHOICE

Catalog No. 11661
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11661 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11661 Supplier Tocris Bioscience™ Supplier No. 1166/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

C-terminal octapeptide of CCK

CCK Octapeptide, sulfated is an endogenous C-terminal octapeptide of CCK found in the central nervous system and gastrointestinal tract.

Non-sulfated Peptide also available.

Specifications

Purity 0.95
Content And Storage Store at -20°C
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.