Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ CGP 42112
GREENER_CHOICE

Catalog No. 25691
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-691 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 25-691 Supplier Tocris Bioscience™ Supplier No. 2569/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective, high affinity AT2 ligand

CGP 42112 is a selective, high affinity angiotensin AT2 receptor ligand (Ki = 0.24 nM). Displays agonistic properties at proximal tubule AT2 receptors, causes Na+, K+-ATPase inhibition and sodium excretion. Antagonizes Ang-II induced contractions in rabbit aortic rings (IC50 = 1850 nM).

Specifications

Purity 95%
Content And Storage Store at -20°C
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.