Learn More
Tocris Bioscience™ Colivelin
GREENER_CHOICE
Catalog No. 3945500U
Change view
Click to view available options
Quantity:
500 μg
Catalog No. Quantity
39-455-00U 500 μg
1 options
This item is not returnable. View return policy
Catalog No. 39-455-00U Supplier Tocris Bioscience™ Supplier No. 3945/500U
Add to Cart
Add to Cart
This item is not returnable. View return policy
STAT3 activator; also protects against β-amyloid neurotoxicity
Colivelin is a neuroprotective peptide and activator of STAT3. Protects neurons against the neurotoxic effects of amyloid β-peptide (1-43) at a concentration of 100 fM in vitro. Suppresses neuronal death by activating STAT3 in vitro; upregulates cholinergic transmission and ameliorates memory impairment in Alzheimer's disease (AD) models. Also prevents alcohol-induced apoptosis in a fetal mouse model. Brain penetrant.
Specifications
Quantity 500 μg