Learn More
Tocris Bioscience™ Compstatin
GREENER_CHOICE
Catalog No. 25851
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
25-851 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 25-851 Supplier Tocris Bioscience™ Supplier No. 2585/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
C3-binding protein, inhibits complement activation
Compstatin is a complement inhibitor; binds C3 inhibiting proteolytic cleavage by C3 convertase (IC50 = 28 μM) and activation of classical and alternative complement pathways (IC50 values are 63 and 12 μM respectively). Prolongs graft survival and reduces hyperacute rejection of ex vivo pig kidneys perfused with human blood and inhibits in vivo heparin/protamine-induced complement activation in primates.
Control Peptide also available.
Specifications
Purity 0.95
Quantity 1 mg