Learn More
Tocris Bioscience™ Cyclotraxin B
GREENER_CHOICE
Catalog No. 50621
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
50621 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 50621 Supplier Tocris Bioscience™ Supplier No. 5062/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
TrkB receptor antagonist
Cyclotraxin B is an antagonist of TrkB receptors; inhibits BDNF-induced TrkB activity (IC50 = 0.30 nM). Allosterically alters TrkB receptor conformation but does not alter BDNF binding. Prevents BDNF-induced cold allodynia in mice. Also shown to exhibit putative anxiolytic properties in mice.
Specifications
Quantity 1 mg