Learn More
Tocris Bioscience™ (d(CH2)51,Tyr(Me)2,Arg8)-Vasopressin
GREENER_CHOICESDP
Catalog No. 33771 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
33-771 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 33-771 Supplier Tocris Bioscience™ Supplier No. 3377/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selective vasopressin V1A antagonist
(d(CH2)51,Tyr(Me)2,Arg8)-Vasopressin is a selective vasopressin V1A receptor antagonist. Inhibits vasopressin and oxytocin-induced increases in intracellular calcium concentrations in vitro (IC50 values are 5 and 30 nM respectively). Exhibits potent and prolonged antivasopressor activity and induces anxiolytic-like effects in the dorsal, but not ventral, hippocampus in vivo.
Specifications
Purity 0.95
Quantity 1 mg