Learn More
Tocris Bioscience™ [D-Lys3]-GHRP-6
GREENER_CHOICE
Catalog No. 19225
Change view
Click to view available options
Quantity:
5 mg
Catalog No. Quantity
19-225 5 mg
1 options
This item is not returnable. View return policy
Catalog No. 19-225 Supplier Tocris Bioscience™ Supplier No. 1922/5
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ghrelin receptor antagonist
[D-Lys3]-GHRP-6 is an antagonist at the ghrelin receptor (GHS-R1a) (IC50 = 0.9 μM). Also weakly binds to melanocortin receptors (Ki = 26-120 μM). Centrally active in vivo.
Specifications
Quantity 5 mg