Learn More
Tocris Bioscience™ [D-p-Cl-Phe6,Leu17]-VIP
GREENER_CHOICE
Catalog No. 30541
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
30-541 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 30-541 Supplier Tocris Bioscience™ Supplier No. 3054/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selective VIP receptor antagonist
[D-p-Cl-Phe6,Leu17]-VIP is a selective vasoactive intestinal peptide (VIP) receptor antagonist (IC50 = 125.8 nM). Displays no activity on glucagon, secretin or GRF receptors.
Specifications
Quantity 1 mg