Learn More
Tocris Bioscience™ Decanoyl-RVKR-CMK
GREENER_CHOICE
Catalog No. 35011
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
35-011 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 35-011 Supplier Tocris Bioscience™ Supplier No. 3501/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Subtilisin/Kex2p-like proprotein convertase inhibitor
Decanoyl-RVKR-CMK is a subtilisin/Kex2p-like proprotein convertase inhibitor; blocks activity of all seven convertases (PC1, PC2, PC4, PACE4, PC5, PC7 and furin). Abolishes proET-1 processing in endothelial cells; inhibits regulated secretion of the neuronal polypeptide VGF in PC12 cells. Inhibits cleavage of glycoprotein B of human cytomegalovirus. Also inhibits cleavage of SARS-CoV-2 spike protein by furin and blocks viral cell entry (IC50 = 57 nM in plaque reduction assay).
Specifications
Quantity 1 mg