Learn More
Tocris Bioscience™ Desmopressin
GREENER_CHOICE
Catalog No. 33961
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
33-961 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 33-961 Supplier Tocris Bioscience™ Supplier No. 3396/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Synthetic vasopressin analog; V1B and V2 agonist
Desmopressin is a synthetic vasopressin analog that acts as an agonist at V1B and V2 receptors (EC50 values are 11.4 and 23.9 nM and Ki values are 5.84 and 65.9 nM respectively). Prevents polycystic kidney disease formation and exhibits antidiuretic, antiproliferative, hemostatic and hypotensive activity in vivo.
Specifications
Purity 0.95
Quantity 1 mg