Learn More
Tocris Bioscience™ Dynamin inhibitory peptide, myristoylated
Description
Control Peptide also available.
Specifications
Specifications
| Purity | 0.95 |
| Common Name | Dynamin Inhibitors |
| Content And Storage | Store at -20°C |
| Quantity | 1 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.