Learn More
Tocris Bioscience™ Endothelin 1 (human, porcine)
GREENER_CHOICE
Catalog No. 1160100U
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
1160100U 100 μg
1 options
This item is not returnable. View return policy
Catalog No. 1160100U Supplier Tocris Bioscience™ Supplier No. 1160/100U
Add to Cart
Add to Cart
This item is not returnable. View return policy
Potent vasoconstrictor peptide
Endothelin 1 (human, porcine) is an endogenous potent vasoconstrictor peptide that is an agonist at ETA and ETB receptors. Modulates vascular tone.