Learn More
Tocris Bioscience™ Exendin-3 (9-39) amide
GREENER_CHOICE
Catalog No. 20811
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
20-811 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 20-811 Supplier Tocris Bioscience™ Supplier No. 2081/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Potent GLP-1 receptor antagonist
Exendin-3 (9-39) amide is a potent and selective GLP-1 receptor antagonist (Kd = 1.7 nM at cloned human GLP-1 receptors). Inhibits cAMP production and insulin release induced by GLP-1 (7-36), exendin-3 (IC50 = 20 nM) and exendin-4. Blocks the inhibitory effect of GLP-1 on food intake in rats.
Specifications
Quantity 1 mg