Learn More
R&D Systems™ Exendin-4
GREENER_CHOICESDP
Catalog No. 19331 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
19-331 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 19-331 Supplier R&D Systems™ Supplier No. 1933/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Potent GLP-1 receptor agonist
Exendin-4 is a high affinity glucagon-like peptide 1 (GLP-1) receptor agonist (Kd = 136 pM); originally isolated from Heloderma suspectum venom. Exendin-4 potently induces cAMP formation without stimulating amylase release in pancreatic acini. Potentiates glucose-induced insulin secretion in isolated rat islets. Exendin-4 protects against glutamate-induced neurotoxicity, and in a mouse model of metabolic imblance it reduces neuroinflammation and enhances long-term-potentiation.
Specifications
Product Line Tocris
Quantity 1 mg