Learn More
Tocris Bioscience™ FSLLRY-NH2
GREENER_CHOICE
Catalog No. 47511
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
47-511 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 47-511 Supplier Tocris Bioscience™ Supplier No. 4751/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
PAR2 peptide antagonist
FSLLRY-NH2 is a selective PAR2 peptide antagonist. Reverses taxol -induced mechanical allodynia, heat hyperalgesia and PKC activation in ICR mice. Blocks ERK activation and collagen production in isolated cardiac fibroblasts. Also reduces symptoms in a mouse model of dermatophyte-associated itch.
Specifications
Purity 0.95
Quantity 1 mg