Learn More
Tocris Bioscience™ Galanin (2-29) (rat)
Catalog No. 14511
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
14-511 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 14-511 Supplier Tocris Bioscience™ Supplier No. 1451/1
This item is not returnable. View return policy
Selective GAL2 peptide agonist
Peptide agonist for galanin receptors, showing selectivity for GAL2 (Ki values are 85, 1.9 and 12.6 nM at rat GAL1, GAL2 and GAL3 receptors respectively).
Specifications
Quantity 1 mg