Learn More
Tocris Bioscience™ Gap 26
Catalog No. 19501
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
19-501 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 19-501 Supplier Tocris Bioscience™ Supplier No. 1950/1
This item is not returnable. View return policy
Gap junction blocker; inhibits smooth muscle contraction and IP3-mediated ATP release
Gap 26 is a peptide, corresponding to residues 63 - 75 of connexin 43, which is a gap junction blocker. Attenuates rhythmic contractile activity of rabbit arterial smooth muscle (IC50 = 28.4 μM). Also inhibits IP3-induced ATP release, without inhibiting gap junctional coupling in endothelial cells.
Specifications
Quantity 1 mg