Learn More
R&D Systems™ Gastrin I (human)
SDP
Catalog No. 30061 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
30-061 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 30-061 Supplier R&D Systems™ Supplier No. 3006/1
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Selective CCK2 agonist
Gastrin I (human) is an endogenous peptide produced in the stomach that acts as a selective CCK2 receptor agonist. Gastrin I stimulates gastric acid secretion and has a mitogenic effect on gastric cells. Gastrin stimulates gastric epithelial cell proliferation and histamine secretion (EC50 values are 6.2 pM and 0.014 nM, respectively). Gastrin I is also used in the culture of stomach, intestine and pancreas organoids. For more information about using Gastrin I in the generation of gastric organoids see the Human Gastric Organoid Culture Protocol .
Specifications
Product Line Tocris
Quantity 1 mg