Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Ghrelin (rat)
GREENER_CHOICE

Catalog No. 14651
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-651 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-651 Supplier Tocris Bioscience™ Supplier No. 1465/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Endogenous ghrelin receptor agonist

Ghrelin (rat) is an endogenous agonist peptide for the ghrelin receptor (GHS-R1a). Produced mainly by the stomach, Ghrelinstimulates release of growth hormone from the pituitary gland in vitro and in vivo, and regulates feeding, growth and energy production.

Specifications

Purity 0.95
Content And Storage Store at -20°C
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.