Learn More
Tocris Bioscience™ GIP (1-39)
GREENER_CHOICE
Catalog No. 22571
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
22-571 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 22-571 Supplier Tocris Bioscience™ Supplier No. 2257/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Highly potent insulinotropic peptide; GIP agonist
GIP (1-39) is an endogenous truncated form of the incretin hormone GIP. More potent at stimulating glucose-dependent insulin secretion from rat pancreatic β-cells than GIP (Cat. No 2084).
Specifications
Quantity 1 mg