Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Glucagon-like peptide 1 (1-37) (human, rat)
GREENER_CHOICE

Catalog No. 18511
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-511 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-511 Supplier Tocris Bioscience™ Supplier No. 1851/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Endogenous pancreatic peptide

Glucagon-like peptide 1 (1-37) (human, rat) is a pancreatic hormone synthesized by post-translational processing of proglucagon. Unlike truncated forms of GLP-1, it has no effect on food intake in rats and does not enhance pancreatic insulin secretion. However it induces insulin expression in intestinal epithelial cells, which can restore glucose homeostasis when implanted into diabetic mice.

Specifications

Purity 95%
Content And Storage Store at -20°C
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.