Learn More
Tocris Bioscience™ GRK2i
Catalog No. 35941
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
35-941 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 35-941 Supplier Tocris Bioscience™ Supplier No. 3594/1
This item is not returnable. View return policy
GRK2 inhibitory polypeptide; Gβ/γ antagonist
GRK2i is a GRK2 inhibitory polypeptide that specifically inhibits Gβγ activation of GRK2. Corresponds to the Gβγ-binding domain and acts as a cellular Gβγ antagonist.
Specifications
Purity 0.95
Quantity 1 mg