Learn More
Tocris Bioscience™ GsMTx4
GSA_VAGREENER_CHOICESDP
Catalog No. 4912100U Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
49-121-00U 100 μg
1 options
This item is not returnable. View return policy
Catalog No. 49-121-00U Supplier Tocris Bioscience™ Supplier No. 4912/100U
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
TRPC1 and TRPC6 inhibitor; inhibits mechanosensitive ion channels
GsMTx4 is a spider venom peptide that selectively inhibits cationic mechanosensitive channels (MSCs) such as TRPC1 and TRPC6 and Piezo channels. GsMTx4 blocks stretch-activated cation channels in astrocytes, cardiac cells, and smooth and skeletal muscle cells. GsMTx4 also inhibits TACAN, a mechanosensitive ion channel involved in the pain response. GsMTx4 decreases the leptin-induced AMPK and MLC-2 phosphorylation in breast epithelial cells. GsMTx4 is neuroprotective and inhibits lysophosphatidylcholine-induced astrocyte toxicity in vivo in mice. GsMTx4 suppresses neurogenesis and enhances astrogenesis in human neural stem cells.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
CAS 1209500-46-8