Learn More
Tocris Bioscience™ Guanylin (human)
GREENER_CHOICE
Catalog No. 1898500U
Change view
Click to view available options
Quantity:
500 μg
Catalog No. Quantity
18-985-00U 500 μg
1 options
This item is not returnable. View return policy
Catalog No. 18-985-00U Supplier Tocris Bioscience™ Supplier No. 1898/500U
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Endogenous activator of intestinal guanylyl cyclase
Guanylin (human) is an endogenous peptide activator of intestinal guanylyl cyclase; secreted mainly by the gastrointestinal mucosa. Regulates water and electrolyte transport in intestinal and renal epithelia.
Specifications
Quantity 500 μg