Learn More
Tocris Bioscience™ Hemokinin 1 (human)
Catalog No. 25761
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
25-761 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 25-761 Supplier Tocris Bioscience™ Supplier No. 2576/1
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Endogenous, selective NK1 agonist
Hemokinin 1 (human) is an endogenous substance P homolog that is a selective agonist at the tachykinin NK1 receptor (IC50 values are 1.8, 370 and 480 nM for NK1, NK3 and NK2 receptors respectively). Has proliferative and antiapoptotic actions on B-cells in vitro and is antihypertensive in vivo.
Specifications
Quantity 1 mg