Learn More
Tocris Bioscience™ Hemopressin (human, mouse)
GREENER_CHOICE
Catalog No. 37911
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
37-911 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 37-911 Supplier Tocris Bioscience™ Supplier No. 3791/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selective CB1 inverse agonist
Hemopressin (human, mouse) is a bioactive endogenous peptide substrate for endopeptidase 24.15 (ep24.15), neurolysin (ep24.16) and ACE. Ki values are 27.76, 3.43 and 1.87 μM respectively. Potent hypotensive in vivo. Also acts as a selective CB1 receptor inverse agonist. Displays antinociceptive activity in vivo.
Specifications
Quantity 1 mg