Learn More
Tocris Bioscience™ HOE 140
GREENER_CHOICE
Catalog No. 3014500U
Change view
Click to view available options
Quantity:
500 μg
Catalog No. Quantity
30-145-00U 500 μg
1 options
This item is not returnable. View return policy
Catalog No. 30-145-00U Supplier Tocris Bioscience™ Supplier No. 3014/500U
Add to Cart
Add to Cart
This item is not returnable. View return policy
Potent and selective B2 antagonist
HOE 140 is a potent and selective bradykinin B2 receptor antagonist (pA2 = 9.04). Also inhibits aminopeptidase N (Ki = 9.1 μM).
Specifications
Purity 0.95
Quantity 500 μg