Learn More
Tocris Bioscience™ Jingzhaotoxin III
GREENER_CHOICE
Catalog No. 4913100U
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
49-131-00U 100 μg
1 options
This item is not returnable. View return policy
Catalog No. 49-131-00U Supplier Tocris Bioscience™ Supplier No. 4913/100U
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selective NaV1.5 channel blocker
Jingzhaotoxin III is a selective blocker of NaV1.5 channels (IC50 = 348 nM); displays no effect on other isoforms, including NaV1.2, NaV1.4, NaV1.6 and NaV1.7. Thought to inhibit sodium channel activation by binding to the NaV1.5 S3-S4 linker of domain II. Selectively inhibits the activation of cardiac sodium channels, but has no effect on sodium channels in dorsal root ganglion neurons.
Specifications
Purity 0.95