Learn More
Tocris Bioscience™ JMV 390-1
Catalog No. 25751
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
25-751 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 25-751 Supplier Tocris Bioscience™ Supplier No. 2575/1
This item is not returnable. View return policy
Inhibits multiple neurotensin and neuromedin N degrading enzymes
Inhibitor of the major neurotensin and neuromedin N degrading enzymes (IC50 values are 31, 40, 52 and 58 nM for endopeptidase 24.15, endopeptidase 24.11, leucine aminopeptidase and endopeptidase 24.16 respectively). Exhibits analgesic effects in various nociception assays.
Specifications
Quantity 1 mg