Learn More
Tocris Bioscience™ Kisspeptin 10 (rat)
GSA_VAGREENER_CHOICE
Catalog No. 42431
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
42-431 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 42-431 Supplier Tocris Bioscience™ Supplier No. 4243/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Endogenous ligand for kisspeptin receptor
Kisspeptin 10 (rat) is an endogenous ligand for the rodent kisspeptin receptor (KISS1, GPR54).
Human Analog also available.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Purity 95%
Quantity 1 mg