Learn More
Tocris Bioscience™ Kisspeptin 234
GREENER_CHOICE
Catalog No. 38811
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
38-811 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 38-811 Supplier Tocris Bioscience™ Supplier No. 3881/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Kisspeptin receptor antagonist; kisspeptin 10 (Cat. No. 2570) analog
Kisspeptin 234 is a kisspeptin receptor (KISS1, GPR54) antagonist; kisspeptin-10 (Cat. No. 2570) analog. Inhibits kisspeptin-10 stimulation of inositol phosphate (IP) (IC50 = 7 nM) and release of gonadotrophin-releasing hormone (GnRH).
Specifications
Quantity 1 mg