Learn More
Tocris Bioscience™ M617
GREENER_CHOICE
Catalog No. 26971
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
26-971 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 26-971 Supplier Tocris Bioscience™ Supplier No. 2697/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selective GAL1 agonist
M617 is a selective galanin GAL1 receptor agonist (Ki values are 0.23 and 5.71 nM for GAL1 and GAL2 receptors respectively). Enhances food consumption in rats following i.c.v. administration and reduces CAP-induced inflammatory pain.
Specifications
Quantity 1 mg