Learn More
Tocris Bioscience™ Margatoxin
Catalog No. 3563100U
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
35-631-00U 100 μg
1 options
This item is not returnable. View return policy
Catalog No. 35-631-00U Supplier Tocris Bioscience™ Supplier No. 3563/100U
This item is not returnable. View return policy
Potent KV1.3 channel blocker
Margatoxin is a potent KV1.3 channel blocker (IC50 = 36 pM). Displays no effect at calcium-activated channels. Reduces VEGF-induced transmembrane calcium influxes and nitric oxide production in human endothelial cells.