Learn More
Tocris Bioscience™ MCH (human, mouse, rat)
GREENER_CHOICE
Catalog No. 3806100U
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
38-061-00U 100 μg
1 options
This item is not returnable. View return policy
Catalog No. 38-061-00U Supplier Tocris Bioscience™ Supplier No. 3806/100U
Add to Cart
Add to Cart
This item is not returnable. View return policy
Potent endogenous MCH agonist
MCH (human, mouse, rat) is a potent endogenous agonist at melanin-concentration hormone (MCH) receptors (IC50 values are 0.3 and 1.5 nM and EC50 values are 3.9 and 0.1 nM at MCH1 and MCH2 receptors respectively). Increases food intake in vivo.