Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Melittin
GREENER_CHOICE

Catalog No. 1193500U
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11935-00U 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11935-00U Supplier Tocris Bioscience™ Supplier No. 1193/500U
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibits Gs and stimulates Gi activity

Melittin is a inhibits Gs and stimulates Gi activity. Inhibits protein kinase C and cAMP-dependent protein kinases. Na+/K+ ATPase inhibitor.

Specifications

Purity 0.95
Content And Storage Store at -20°C
Quantity 500 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.