Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Motilin (human, porcine)

Catalog No. 22641
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-641 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-641 Supplier Tocris Bioscience™ Supplier No. 2264/1
This item is not returnable. View return policy

Endogenous peptide regulator of gastrointestinal motility

Motilin (human, porcine) is an endogenous motilin receptor ligand (Ki = 2.3 nM) that regulates gastrointestinal motor function. Stimulates contraction of gut smooth muscle (EC50 = 1 nM) and increases pepsin release.

Specifications

Purity 95%
Content And Storage Store at -20°C
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.