Learn More
Tocris Bioscience™ Myelin Basic Protein (87-99)
Catalog No. 33991
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
33-991 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 33-991 Supplier Tocris Bioscience™ Supplier No. 3399/1
This item is not returnable. View return policy
Major antigenic component implicated in the pathophysiology of multiple sclerosis
Myelin Basic Protein (87-99) is an encephalitogenic peptide that induces T cell proliferation with Th1 polarization in the CNS. Major antigenic component implicated in the pathophysiology of multiple sclerosis.
Specifications
Purity 0.95
Quantity 1 mg