Learn More
Tocris Bioscience™ Neuropeptide Y (human, rat)
GREENER_CHOICE
Catalog No. 1153200U
Change view
Click to view available options
Quantity:
200 μg
Catalog No. Quantity
11532-00U 200 μg
1 options
This item is not returnable. View return policy
Catalog No. 11532-00U Supplier Tocris Bioscience™ Supplier No. 1153/200U
Add to Cart
Add to Cart
This item is not returnable. View return policy
Influences feeding and sexual behavior
Neuropeptide Y (human, rat) is a widely distributed endogenous neuropeptide involved in the control of food intake, sexual behavior and blood pressure.
Specifications
Quantity 200 μg