Learn More
Tocris Bioscience™ Neurotensin
GREENER_CHOICE
Catalog No. 19091
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
19-091 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 19-091 Supplier Tocris Bioscience™ Supplier No. 1909/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Endogenous neurotransmitter and endocrine modulator
Neurotensin is a brain and gastrointestinal peptide with many central and peripheral functions. Acts as neuromodulator of dopamine transmission and exerts potent hypothermic and analgesic effects. Peripherally, acts as a paracrine and endocrine modulator of the digestive tract and cardiovascular system.
Specifications
Quantity 1 mg