Learn More
Tocris Bioscience™ NFAT Inhibitor
GREENER_CHOICE
Catalog No. 39301
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
39-301 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 39-301 Supplier Tocris Bioscience™ Supplier No. 3930/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Inhibitor of calcineurin-mediated NFAT activation
NFAT Inhibitor is a selective inhibitor of calcineurin-mediated dephosphorylation of nuclear factor of activated T cells (NFAT). Does not disrupt other calcineurin-dependent pathways. Inhibits NFAT activation and NFAT-dependent expression of endogenous cytokine genes in T cells.
Specifications
Purity 0.95
Quantity 1 mg